Order Entry
ContactUsLinkComponent
Compact Dry E. Coli (EC), 240 Plates per Pack, ready-to-use test method, for determination of total aerobic bacterial counts, contains two kinds of chromogenic enzyme substrates: Magenta-Gal & X-Gluc, E. Coli forms blue colonies, coliforms turn red, and sum of both will be total coliform group count
CompactDry™ E. coli (EC), Hardy Diagnostics
Catalog #: 76414-990
CAS Number:  
Compact Dry E. Coli (EC), 240 Plates per Pack, ready-to-use test method, for determination of total aerobic bacterial counts, contains two kinds of chromogenic enzyme substrates: Magenta-Gal & X-Gluc, E. Coli forms blue colonies, coliforms turn red, and sum of both will be total coliform group count
CompactDry™ E. coli (EC), Hardy Diagnostics
Catalog #: 76414-990
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Packaging:
    240 plates per pack
  • Description:
    CompactDry™ E. coli (EC) and coliforms cassettes for colony counting
  • Storage temperature:
    2 to 30 °C away from direct light
  • Cat. no.:
    76414-990
  • Includes:
    25 Packs

Specifications

About this item

CompactDry™ EC is a ready-to-use test method recommended for the isolation and enumeration of coliforms and Escherichia coli in raw materials, finished products, or on environmental surfaces pertaining to food and related industries.

  • Sturdy frame, stackable packaging
  • Self-diffusing media
  • Ability to subculture without damage to colonies
  • No leakage
  • Easy to label

CompactDry™ EC contains two kinds of chromogenic enzyme substrates: Magenta-Gal and X-Gluc. E. coli forms blue colonies, coliforms turn pink-purple. The total coliform group count is the sum of both the pink-purple and blue colonies.

Each cassette comes with 4 connected plates that can be easily snapped apart to support a variety of dilution scenarios.

Caution: Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.
Customers Who Bought This Also Bought