Copper wire coil, open core 22 gauge, consist of 220 turns of enamel coated magnet wire, 2x1inch by 1/4inch thick, weigh about 1 ounce or [28 grams] and have a resistance of approximately 8 ohms, pack of 4
Catalog #: 470326-952
Supplier: PICOSOLUTIONS GROUP INC SE
Customers Who Bought This Also Bought