ContactUsLinkComponent
Cryo Rack, 48 Positions, with predefined 1D and 2D barcodes, with lid, translucent, Polypropylene, Color: Black, Pre-Coded Digit, Low Profile, CODE 128, S1 Centered, Human readable number includes Datamatrix code on bottom of the rack, non-sterile, for 1ml and 2ml cryo tubes
Catalog #: 76414-146
Supplier:  Greiner Bio-One
Cryo Rack, 48 Positions, with predefined 1D and 2D barcodes, with lid, translucent, Polypropylene, Color: Black, Pre-Coded Digit, Low Profile, CODE 128, S1 Centered, Human readable number includes Datamatrix code on bottom of the rack, non-sterile, for 1ml and 2ml cryo tubes
Catalog #: 76414-146
Supplier:  Greiner Bio-One
Customers Who Bought This Also Bought