ContactUsLinkComponent
VCV 1 Vacuum control valve manual, Allows for adjusting the set vacuum value on the manual manometer, Rod and stand Not included in delivery
Catalog #: 76414-132
Supplier:  IKA Works
VCV 1 Vacuum control valve manual, Allows for adjusting the set vacuum value on the manual manometer, Rod and stand Not included in delivery
Catalog #: 76414-132
Supplier:  IKA Works
Customers Who Bought This Also Bought