Samples tongs, for MA37/MA160, For easy and secure handling of aluminum sample pans
Catalog #: CA76413-054
Supplier: Sartorius
Samples tongs, for MA37/MA160, For easy and secure handling of aluminum sample pans
Catalog #: CA76413-054
Supplier: Sartorius
Customers Who Bought This Also Bought
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with Certificate of Calibration, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$31.17 to $282.75
VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Supplier: VWR International
$69.94