ContactUsLinkComponent
Holder, for Weighing Boats or Papers, allows easy positioning and secure fixation of weighing boats or weighing papers on the pan. Moreover, also reduces influences of electrostatic influences because Faraday Cage Effect.
Catalog #: CA76413-036
Supplier:  Sartorius
Holder, for Weighing Boats or Papers, allows easy positioning and secure fixation of weighing boats or weighing papers on the pan. Moreover, also reduces influences of electrostatic influences because Faraday Cage Effect.
Catalog #: CA76413-036
Supplier:  Sartorius
Customers Who Bought This Also Bought