ContactUsLinkComponent
Cal-Pak* Weight set, ASTM Class 1 with NVLAP Certificate, Contains three high quality precision weights 200G 20G 20MG, Can be used with all balance, Supplied in a rugged polycarbonate case with each individual weight in its own high quality polycarbonate case
Catalog #: CA76412-514
Supplier:  Sartorius
Cal-Pak* Weight set, ASTM Class 1 with NVLAP Certificate, Contains three high quality precision weights 200G 20G 20MG, Can be used with all balance, Supplied in a rugged polycarbonate case with each individual weight in its own high quality polycarbonate case
Catalog #: CA76412-514
Supplier:  Sartorius
Customers Who Bought This Also Bought