ContactUsLinkComponent
Cal-Pak* Weight set, ASTM Class 0 with NVLAP Certificate, Contains three high quality precision weights 2G 200MG 1MG, Can be used with all balance, Supplied in a rugged polycarbonate case with each individual weight in its own high quality polycarbonate case
Catalog #: CA76412-486
Supplier:  Sartorius
Cal-Pak* Weight set, ASTM Class 0 with NVLAP Certificate, Contains three high quality precision weights 2G 200MG 1MG, Can be used with all balance, Supplied in a rugged polycarbonate case with each individual weight in its own high quality polycarbonate case
Catalog #: CA76412-486
Supplier:  Sartorius
Customers Who Bought This Also Bought