8-425 Screw-Thread Vial Caps
Catalog #: 76414-878
Supplier: Thermo Scientific
Specifications
- Cap Size:8 mm
- Cap Type:Polypropylene
- Septum:None
- Description:Closed top screw cap, not magnetic, 8 mm
- Cap Color:Black
Specifications
About this item
Customers Who Bought This Also Bought
Chloroform ≥99.8% (chloroform + ethanol basis) stabilized, GR ACS, Supelco®
Supplier: MilliporeSigma
$981.40 to $3,925.60
Water ACS, Reagent Grade ASTM Type I, ASTM Type II Packaged in plastic containers
Supplier: Ricca Chemical
$200.03
Sodium thiosulfate 0.10 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$153.23 to $602.78
Masterflex® L/S® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
$276.50
Ethanol denatured ACS (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Supplier: VWR International
$411.94 to $1,303.38
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Citric acid monohydrate 99.5-100.5% (by acidimetry on anhydrous substance), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
$365.38 to $2,062.18
Sodium bisulfite ≥90% FCC, purified, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$237.49 to $1,424.99
Clarity Urocheck Urinalysis Strips, Clarity Diagnostics
Supplier: Clarity Diagnostics
$31.63 to $756.61