Stand up Holder for Trio Bas Mono, Duo, and Trio Air Sampler, Fabricated with 100% high density impact resistant technopolymer, The shaped base allows the vertical positioning of the air sampler with the use of just one hand, Designed to position on all tripods and MINI multi holder cart with wheels
Catalog #: CA103792-978
Supplier: HARDY DIAGNOSTICS CA
Customers Who Bought This Also Bought