ContactUsLinkComponent
ProSwap Cable Assembly, With shallow Depth, with temperature and depth sensors, features are accommodate Optical Dissolved Oxygen, pH, Total Algae-PC (Phycocyanin + Chlorophyll), Total AlgaePE (Phycoerythrin + Chlorophyll), Ammonium ISE, Nitrate ISE, Chloride ISE, PH/ORP, not compatible, Size: 50m
Catalog #: 76411-476
Supplier:  YSI
ProSwap Cable Assembly, With shallow Depth, with temperature and depth sensors, features are accommodate Optical Dissolved Oxygen, pH, Total Algae-PC (Phycocyanin + Chlorophyll), Total AlgaePE (Phycoerythrin + Chlorophyll), Ammonium ISE, Nitrate ISE, Chloride ISE, PH/ORP, not compatible, Size: 50m
Catalog #: 76411-476
Supplier:  YSI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.