ContactUsLinkComponent
ProSwap cable assembly, no depth, Includes temperature sensor, order other sensor separately, Conductivity/Temperature, features are accommodate Optical Dissolved Oxygen, pH, Total Algae-PC (Phycocyanin + Chlorophyll), Total AlgaePE (Phycoerythrin + Chlorophyll), Ammonium ISE, Nitrate ISE, Size: 10m
Catalog #: 76411-456
Supplier:  YSI
ProSwap cable assembly, no depth, Includes temperature sensor, order other sensor separately, Conductivity/Temperature, features are accommodate Optical Dissolved Oxygen, pH, Total Algae-PC (Phycocyanin + Chlorophyll), Total AlgaePE (Phycoerythrin + Chlorophyll), Ammonium ISE, Nitrate ISE, Size: 10m
Catalog #: 76411-456
Supplier:  YSI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.