ContactUsLinkComponent
ProSwap Digital Water Quality Meter, with gps, Swap one smart sensor for another with the ProSwap digital water quality meter, Featuring built-in temperature and optional depth sensors, ProSwap can transform into whatever you need, purpose-built turbidity meter or a high-accuracy optical DO meter
Catalog #: 76411-450
Supplier:  YSI
ProSwap Digital Water Quality Meter, with gps, Swap one smart sensor for another with the ProSwap digital water quality meter, Featuring built-in temperature and optional depth sensors, ProSwap can transform into whatever you need, purpose-built turbidity meter or a high-accuracy optical DO meter
Catalog #: 76411-450
Supplier:  YSI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.