Order Entry
ContactUsLinkComponent
SALT-T4 DNA LIGASE 20000 UNITS
Salt-T4 DNA Ligase, New England Biolabs
Catalog #: CA76407-346
CAS Number:  
SALT-T4 DNA LIGASE 20000 UNITS
Salt-T4 DNA Ligase, New England Biolabs
Catalog #: CA76407-346
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    400000 units/ml
  • Description:
    Salt-T4 DNA ligase
  • Size:
    20000 units
  • Cat. no.:
    CA76407-346

Specifications

About this item

A salt tolerant variant of T4 DNA Ligase, Salt-T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5 phosphate and 3 hydroxyl termini in duplex DNA or RNA and is designed to function at higher salt concentrations than wild type T4 DNA Ligase.

  • Salt-T4 DNA Ligase is an engineered variant of T4 DNA
  • Ligase with improved salt tolerance
  • Capable of joining blunt end and cohesive end termini
  • Capable of repairing single stranded nicks in duplex DNA and some DNA/RNA hybrids

This enzyme will join cohesive end termini at salt concentrations as high as 300 mM without any loss in activity. This enzyme is insensitive to salt carried over from other reaction components (vector or insert DNA) and allows ligation reactions to proceed in alternative reaction buffers with higher levels of salt (e.g., NEBuffer 3.1).

Delivery: Supplied with both T4 DNA ligase buffer and StickTogether DNA ligase buffer.
Customers Who Bought This Also Bought