ContactUsLinkComponent
EPDM Isolator Gloves, non-sterile, Comfortable and dexterous with great tactile sensitivity, Resistant against hydrogen peroxide solutions and common disinfecting chemicals, Resists aging from exposure to oxygen, UV rays and ozone,Size: 32in long, 24mil thick, 8in port, Ambidextrous, Size: 11
Catalog #: CA76408-490
Supplier:  Ansell Canada
EPDM Isolator Gloves, non-sterile, Comfortable and dexterous with great tactile sensitivity, Resistant against hydrogen peroxide solutions and common disinfecting chemicals, Resists aging from exposure to oxygen, UV rays and ozone,Size: 32in long, 24mil thick, 8in port, Ambidextrous, Size: 11
Catalog #: CA76408-490
Supplier:  Ansell Canada
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.