ContactUsLinkComponent
CSM Isolator Gloves, non-sterile, Designed for high resistance against concentrated acids and bases, Soft, flexible material designed for ease of use, Resists ageing from exposure to oxygen, Withstands temperatures up to 120 degree C, 32in long, 24mil thick, 12in port, Ambidextrous, Size: 9.5
Catalog #: CA76408-480
Supplier:  Ansell Canada
CSM Isolator Gloves, non-sterile, Designed for high resistance against concentrated acids and bases, Soft, flexible material designed for ease of use, Resists ageing from exposure to oxygen, Withstands temperatures up to 120 degree C, 32in long, 24mil thick, 12in port, Ambidextrous, Size: 9.5
Catalog #: CA76408-480
Supplier:  Ansell Canada
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.