ContactUsLinkComponent
CSM Isolator Gloves, non-sterile, Designed for high resistance against concentrated acids and bases, Soft, flexible material designed for ease of use, Resists ageing from exposure to oxygen, Withstands temperatures up to 120 degree C, 32in long, 16mil thick, 10in port, Ambidextrous, Size: 11
Catalog #: CA76408-470
Supplier:  Ansell Healthcare
CSM Isolator Gloves, non-sterile, Designed for high resistance against concentrated acids and bases, Soft, flexible material designed for ease of use, Resists ageing from exposure to oxygen, Withstands temperatures up to 120 degree C, 32in long, 16mil thick, 10in port, Ambidextrous, Size: 11
Catalog #: CA76408-470
Supplier:  Ansell Healthcare
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.