ContactUsLinkComponent
EPDM Isolator Gloves, non-sterile, Comfortable and dexterous with great tactile sensitivity, Resistant against hydrogen peroxide solutions and common disinfecting chemicals, Resists aging from exposure to oxygen, UV rays and ozone,Size: 32in long, 16mil thick, 12in port, Ambidextrous, Size: 9.5
Catalog #: CA76408-500
Supplier:  Ansell Healthcare
EPDM Isolator Gloves, non-sterile, Comfortable and dexterous with great tactile sensitivity, Resistant against hydrogen peroxide solutions and common disinfecting chemicals, Resists aging from exposure to oxygen, UV rays and ozone,Size: 32in long, 16mil thick, 12in port, Ambidextrous, Size: 9.5
Catalog #: CA76408-500
Supplier:  Ansell Healthcare
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.