ContactUsLinkComponent
Temperature/humidity calibration, 9 temperature measuring points and 1 humidity measuring point - including certificate, 9 temperature measuring points and 1 humidity measuring point in center of chamber, at 25 degreeC / 60 % RH or at specified values - for MKF, KMF, KBF, KBF P, KBWF, MKFT Series
Catalog #: 76410-142
Supplier:  Binder
Temperature/humidity calibration, 9 temperature measuring points and 1 humidity measuring point - including certificate, 9 temperature measuring points and 1 humidity measuring point in center of chamber, at 25 degreeC / 60 % RH or at specified values - for MKF, KMF, KBF, KBF P, KBWF, MKFT Series
Temperature/humidity calibration, 9 temperature measuring points and 1 humidity measuring point - including certificate, 9 temperature measuring points and 1 humidity measuring point in center of chamber, at 25 degreeC / 60 % RH or at specified values - for MKF, KMF, KBF, KBF P, KBWF, MKFT Series
Catalog #: 76410-142
Supplier:  Binder
Customers Who Bought This Also Bought