TSX Series High-Performance -20C Manual Defrost Freezer, Right Promo, Door Style: Solid, Shelves: 4, Plug Type: NEMA 5-15, Voltage:115 V 60 Hz, Dimensions (L x W x H) Exterior: 37.5 x 28 x 78.6 in, Size: 23cu.ft
Catalog #: MP75839-182
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought