Sani stick kit Sponge on handle with Letheen broth, Application: collect samples to detect the presence of microbiological on any surface in food, public health, and cosmetic industries, Mil in: 4.0, MICR: 101, Dimension: 6.3x11In, 20oz, 140 x279mm
Catalog #: CA75849-320
Supplier: Labplas
Customers Who Bought This Also Bought