ContactUsLinkComponent
REPLIQA HIFI TOUGHMIX 500 REACTIONS 25UL
repliQa HiFi ToughMix, Quantabio
Catalog #: CA76382-790
Supplier:  Quantabio
CAS Number:  
repliQa HiFi ToughMix, Quantabio
Catalog #: CA76382-790
Supplier:  Quantabio
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    repliQa HiFi ToughMix
  • Size:
    500 reactions
  • Cat. no.:
    CA76382-790
Specifications
About this item
Superior speed and inhibitor tolerance.
  • High fidelity with >90× wild type Taq
  • Up to 3× faster PCR results with extension rates as fast as 1 kb/sec
  • Tough tested and tolerant to a wide range of PCR inhibitors
  • Superior sensitivity means higher yields from lower inputs as little as 2 pg
  • Has the ability to amplify long fragments of genomic DNA (+24 kb) and lambda DNA (+40 kb)
The repliQa HiFi ToughMix is a 2×, ready-to-use master mix that contains all the components for high fidelity PCR amplification, including a genetically modified DNA polymerase coupled with hot start antibodies. This unique, next generation master mix provides 90× higher fidelity compared to Taq, while reducing time to PCR results by 2 to 3×. The extreme speed is enabled by extension times as fast as 1 to 10 kb/sec depending on target length. The enzyme is coupled with the industry leading ToughMix which is tolerant to a wide variety of inhibitors making it suitable for routine PCR, cloning, amplicon sequencing and site directed mutagenesis.
This mix also has fast extension times, which significantly shortens time to results. Significantly faster results (up to 3×) with 1 kb/sec extension rate for fragments ≤1 kb with no sacrifice in quality or yield of PCR results.
Tolerant to a wide range of PCR inhibitors, it is able to tolerate a wide range of common PCR inhibitors, such as ethanol, hemoglobin and xylan, allowing for amplification of crude or difficult PCR sample types. Can amplify from crude samples.
It provides higher yield and sensitivity, highlighting the enzyme's high efficiency. Coupled with extreme amplification speed, PCR products are able to be detected earlier and at lower levels. Consistently achieve higher yields across a range of inputs.
It is able to amplify varying levels of GC-content targets (32 to 70% GC-rich), further enabling superior PCR performance with bacterial, plant and mammalian sample types.
Customers Who Bought This Also Bought