PIPETMAN® Multichannel Mechanical Pipettor, Gilson
Catalog #: 76408-366
Supplier: GILSON, INC.
Specifications
- Accuracy:±0.50 - ±2.00 µl
- Volume:20 - 200 µl
- Description:P12×200
- Imprecision:≤0.25 - ≤0.50 µl
- Cat. no.:76408-366
- For Use With:D200/DF200/D300/DF300 tips
- Color Code:Yellow
Specifications
About this item
Mechanical air-displacement pipette which offers robustness, accuracy and precision as well as a very low pipetting force for the highest comfort.
- Very low pipetting forces thanks to easy glide piston displacement
- Color-coded push button ensures easy identification and GLP compliance
- Made of PVDF and stainless steel
- Equipped with clip ejector for convenient access to the serviceable parts
- Blue ring positioned under the thumbwheel allows to identify PIPETMAN® and to distinguish it from the Classic and Neo models
Pipettes are individually calibrated and delivered with a certificate of conformity. Each pipette bears an engraved identification number for traceability.
Ordering information: Models equipped with a tip ejector are only sold with a stainless steel tip ejector. Plastic tip ejectors are available in the accessories.
Customers Who Bought This Also Bought