ContactUsLinkComponent
Conforming Stretch Gauze, Non-Sterile, Rayon/poly knitted stretch gauze bandage, Specifically designed not to slip while in use, Unique weave construction and finished edges prevent unraveling and linting, Dimensions: 2INX4.1YD
Conforming Stretch Gauze, DUKAL™ Corporation
Catalog #: 76407-576
Supplier:  Dukal
Conforming Stretch Gauze, Non-Sterile, Rayon/poly knitted stretch gauze bandage, Specifically designed not to slip while in use, Unique weave construction and finished edges prevent unraveling and linting, Dimensions: 2INX4.1YD
Conforming Stretch Gauze, DUKAL™ Corporation
Catalog #: 76407-576
Supplier:  Dukal
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Dimensions:
    2" × 4.1 yds.
  • Sterility:
    Non sterile
  • Cat. no.:
    76407-576
Specifications
About this item
Rayon/poly knitted stretch gauze bandage.
  • Specifically designed not to slip while in use
  • Available in non sterile and sterile formats
  • No natural rubber latex
Conforming stretch gauze provides slight compression to the site while securely holding to any body contour. The unique weave construction and finished edges prevent unraveling and linting.
Customers Who Bought This Also Bought