ContactUsLinkComponent
Tubing, APST, translucent, platinum-cured USP Class VI silicone,extruded in a Class 1000 clean room & stored in a particulate-controlled warehouse, highly pure product meets FDA,EP,3A,ISO standards, Dimensions: 0.063 X 0.188 in
Catalog #: 76407-456
Supplier:  NewAge Industries
Tubing, APST, translucent, platinum-cured USP Class VI silicone,extruded in a Class 1000 clean room & stored in a particulate-controlled warehouse, highly pure product meets FDA,EP,3A,ISO standards, Dimensions: 0.063 X 0.188 in
Catalog #: 76407-456
Supplier:  NewAge Industries