Sabdex (Sabouraud Dextrose) Agar, USP, SterEM, irradiated, triple bagged, Dimension: 15x100mm plate, Volume: 26ml fill, for isolation, cultivation, maintenance of non-pathogenic, pathogenic species of fungi, yeast, meet harmonized USP/EP/JP standard for microbial examination of non sterile products
Catalog #: CA76200-288
Supplier: HARDY DIAGNOSTICS CA
Customers Who Bought This Also Bought