Lok-Tight™ Contact Plates for Surface Sampling, Hardy Diagnostics
Catalog #: CA76200-282
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Packaging:15×65 mm CP
- Description:Tryptic Soy Agar (TSA), with Lecithin and Tween® 80, USP, Irradiated, Triple Bagged, Red Tinted Contact Plate
- Cat. no.:CA76200-282
Specifications
About this item
Contact Plates with Friction Lids to ensure sterility in your environmental testing program.
- Tear notch on outer bag for easy opening; no scissors required
- New moisture control packaging format prevents moisture buildup and ensures safe and sterile transfer
- Friction lid design keeps lids in place
- Lok-Tight™ optional locking feature means lids stay secured during sampling and transport
- Each plate has a grid molded into the bottom of the plate to aid in the counting of microorganisms
- Certificate of analysis available online for each lot
These contact plates are recommended for use in the cultivation of microorganisms from environmental surfaces.
Customers Who Bought This Also Bought