ContactUsLinkComponent
Ultra-Low Temperature Upright Freezer, Electrical: 220V, Interior Dimension: 79x88.2x199.3 cm, Exterior Dimensions: 63x60x140 cm, Weight: 246 kg, Reliable, Maximum storage, Minimal footprint, VIP Plus insulation composite,Volume: 528 l (18.6 cu.ft.)
Catalog #: MP76269-676
Supplier:  PHC Corporation
Ultra-Low Temperature Upright Freezer, Electrical: 220V, Interior Dimension: 79x88.2x199.3 cm, Exterior Dimensions: 63x60x140 cm, Weight: 246 kg, Reliable, Maximum storage, Minimal footprint, VIP Plus insulation composite,Volume: 528 l (18.6 cu.ft.)
Catalog #: MP76269-676
Supplier:  PHC Corporation
Customers Who Bought This Also Bought