SW® PowerForm® PF-95GW Dual-Color Green/White 5.9 mil Sustainable Skin-Improving Nitrile Exam Gloves
Catalog #: 76406-616
Supplier: SW Safety Solutions
Minority or Woman-Owned Business Enterprise
Specifications
- Ambidextrous/Hand-specific:Ambidextrous
- Food contact approved:Yes
- Glove Color:Green
- Glove size:M
- Length:242 mm (9.5")
- Powdered/Powder-free:Powder-free
- Disposable / reusable:Disposable
- Latex/Latex-free:Latex-free
- Model:PF-95GW
- Thickness:0.13 mm (4.9 mil)
- EN norm:EN ISO 21420:2020; EN 455-2000-2015
- FDA regulated:+
- ASTM No.:D6319-19; D6978-05 (2019); F1671-13; F739-12
- Cuff style:Beaded
- Finger thickness:0.15 mm (5.9 mil)
- Palm thickness:0.13 mm (4.9 mil)
- Cuff thickness:0.10 mm (3.8 mil)
- Cuff length:242 mm (9.5")
- Tensile strength:18 Mpa
- Ultimate elongation:500%
- Finish:Fully textured
Specifications
About this item
Experience EnerGel® infused gloves, designed to hydrate skin and protect against long-term glove usage, with sustainable EcoTek® technology.
- EnerGel® hand conditioning technology hydrates and soothes skin; proven to prevent many skin ailments
- EcoTek® sustainable technology
- Breach Alert® visual detection technology provides durable 2-ply construction and contrasting inner color
- Proven resistance to fentanyl and gastric acid for over 4 hours to replicate hazardous settings and overdose conditions
- Fully textured for secure grip
- Silicone-free for critical environments
- 1.5 AQL exceeds laboratory standards
EcoTek® gloves tested to ASTM D5526-12 standard test method for determining anaerobic biodegradation of plastic materials under accelerated landfill conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated. For additional information on the test reports, contact [email protected].
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; SHA Dermatologically Approved; EAC TP TC 019/2011; CE 0493; CE EN455.
Ordering information: Glove sizes are as follows; XS (5.5 to 6), S (6.5 to 7), M (7.5 to 8), L (8.5 to 9), XL (9.5 to 10), XXL (10.5 to 11).
Customers Who Bought This Also Bought