NEBNext Companion Module for Oxford Nanopore Technologies Ligation Sequencing, New England Biolabs
Catalog #: CA76339-730
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:NEBNext companion module for Oxford Nanopore technologies ligation sequencing
- No. of Reactions:24
Specifications
About this item
The NEBNext DNA repair, end repair and ligation reagents recommended in Ligation library preparation are now available in the same product, at volumes designed for use in several protocols.
- Component volumes tailored for use with SQK-LSK109
- Simplified ordering and inventory management
- Compatible with all devices: MinION, GridION, PromethION, Flongle
- No more waste - no unnecessary buffers or excess reagents
The appropriate device-specific SQK-LSK109 protocol from Oxford Nanopore Technologies should be followed for use of these reagents.
Delivery: This module contains the following reagents, at volumes designed for use alongside Oxford Nanopore Technologies SQK-LSK109: NEBNext FFPE DNA Repair Mix (0.048 ml), NEBNext FFPE DNA Repair Buffer (0.084 ml), NEBNext Ultra II End Prep Enzyme Mix (0.072 ml), NEBNext Ultra II End Prep Reaction Buffer (0.084 ml) and Quick T4 DNA Ligase (0.240 ml).
Customers Who Bought This Also Bought