Order Entry
ContactUsLinkComponent
Rt Supermix Kit 100Rxn F/Cdna Synthesis
LunaScript RT SuperMix Kits, New England Biolabs
Catalog #: CA76285-188
CAS Number:  
Rt Supermix Kit 100Rxn F/Cdna Synthesis
LunaScript RT SuperMix Kits, New England Biolabs
Catalog #: CA76285-188
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    LunaScript RT SuperMix kit
  • Size:
    100 reactions
  • No. of Reactions:
    100

Specifications

About this item

LunaScript RT SuperMix Kit features an optimized master mix containing all the necessary components for first strand cDNA synthesis in the context of a two-step RT-qPCR workflow.

  • This kit is an optimized master mix for first strand cDNA synthesis in a two-step RT-qPCR workflow
  • Less than 15 minute first-strand cDNA synthesis protocol
  • Novel, thermostable Luna Reverse Transcriptase improves performance
  • Combine with Luna qPCR master mixes for robust RT-qPCR results
  • Enables consistent linearity, sensitivity, and capacity for reliable RNA quantification

It includes the thermostable Luna Reverse Transcriptase, which supports cDNA synthesis at elevated temperatures. Murine RNase Inhibitor is also included to protect template RNA from degradation. The LunaScript RT SuperMix contains random hexamer and poly-dT primers, allowing even coverage across the length of the RNA targets. In addition, the LunaScript RT SuperMix Kit contains a blue dye, providing a visual indicator that can be followed throughout the two-step RT-qPCR process. The LunaScript RT SuperMix offers robust, linear, and sensitive detection using total RNA inputs as high as 1 µg and as low as single copies of RNA.

Customers Who Bought This Also Bought