Avantor® ACE® Guard Column Cartridge Holders
Catalog #: 76395-216
Supplier: HICHROM LIMITED
Specifications
- Description:Semi-preparative cartridge holder for 7.75 to 21.2 mm Int.Ø. columns
- Cat. no.:76395-216
Specifications
About this item
Stainless steel Avantor® ACE® stand-alone and integral guard cartridge holders are used for connecting guard cartridges to the LC column to provide column protection.
Integral holders connect directly to the analytical column. Stand-alone holders require the use of a suitable connector.
Customers Who Bought This Also Bought