Avantor® Hichrom, Chromatography Tubing, Stainless Steel
Catalog #: 76397-948
Supplier: HICHROM LIMITED
Specifications
- Ø:1.5 mm
- I.D.:1.00 mm
- Length:1.5 m
- Cat. no.:76397-948
Specifications
About this item
Tubings are madeup of 316 Stainless steel, precision bore ¹/₁₆" type tubing.
- Operating temperature - no limit
- May corrode with chloride salt or other buffers
- May adsorb proteins
These 0.005 and 0.007" Ø - suitable for minimal dead volume injector-column or column-detector connections of microbore or short columns containing 3 µm material. Sample and material must be filtered to avoid blockage.
The tube with 0.10' Ø - suitable for connection of analytical and semi-preparative columns containing 5 and 10 µm material. It should be used in all parts of the system where dead volume needs to be kept to minimum.
These 0.20, 0.30 and 0.40" Ø - suitable for use in preparative systems or in non-critical connections (pump to sample injector where low flow resistance is more important than low internal volume).
Customers Who Bought This Also Bought