Biohazard Waste Bags, Autoclavable, Medegen Medical Products
Catalog #: 76380-286
Supplier: Medegen Medical Products, LLC
Specifications
- Capacity:13 - 16 gal.
- Color:Red
- Thickness:1.8 mil
- Cat. no.:76380-286
- Closure:Twist tie
Specifications
About this item
These autoclave bags are built to withstand the rigors of steam sterilization while meeting the requirements for the collection, storage, transport and processing of biohazardous waste.
- Standard autoclave bags are tested to exceed 165 g of dart impact for compliance in the majority of US states
- Biohazardous labeling in both English and Spanish
- International Biohazard Symbol in easy-to-see contrasting color
Delivery: Twist ties included.
Packaging: Flat pack.
Caution: Not for use in CA, FL or PA.
Customers Who Bought This Also Bought