Order Entry
ContactUsLinkComponent
Powder Thief Sterileware 100Cm 20Ml PK20
SP Bel-Art® Sterileware® Powder Thief, Bel-Art Products, a part of SP
Catalog #: 76402-920
Powder Thief Sterileware 100Cm 20Ml PK20
SP Bel-Art® Sterileware® Powder Thief, Bel-Art Products, a part of SP
Catalog #: 76402-920

Specifications

  • Capacity:
    20 ml (0.67 oz.)
  • Length:
    100 cm (39⅜")
  • Cat. no.:
    76402-920

Specifications

About this item

These sterile powder thieves easily sample from targeted locations within bulk powders. Used by pharmaceutical, food, cosmetic and chemical companies.

  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) thief heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10⁻⁶)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization

Available in two lengths each with four tip volumes.

Ordering information: Sterilized products are not returnable.
Customers Who Bought This Also Bought