Order Entry
ContactUsLinkComponent
Vwr Pbs Buffer Powder Pack 1X 1L CS100
VWR® PBS (Phosphate Buffered Saline) Buffer Powder Packs
Catalog #: 76371-734
Supplier:  VWR International
CAS Number:  
Vwr Pbs Buffer Powder Pack 1X 1L CS100
VWR® PBS (Phosphate Buffered Saline) Buffer Powder Packs
Catalog #: 76371-734
Supplier:  VWR International
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • pH:
    7.2 - 7.6
  • Description:
    VWR® PBS Buffer Powder
  • Size:
    100 pks
  • Formulation:
    137 mM NaCl, 2.7 mM KCl, 10mM phosphate

Specifications

About this item

VWR® Buffer Powder Packs can save time and money, as there is no need to measure and weight many separate chemical components which is required when making buffers from scratch.

  • Buffer powder quickly dissolves to make 1L of 1X solution
  • No weighing or measuring of separate components
  • No pH adjustments required
  • Economical shipping, long shelf life
  • Quality and consistency guaranteed

VWR® Pre-measured buffer packs are extremely easy to use and are formulated so each pack makes 1 Liter of 1X buffer. Simply tear open a pack and mix completely with 1 liter of distilled water.

Shipping costs are also minimal, as each case of 50 or 100 packs is compact and weighs only 2.5 Lbs.

Certifications: COA avaialble for each lot number
Packaging: Each case of buffer packs is packaged in a convenient dispenser box, which takes up very little lab space. Included in each case is an instruction insert, and a roll of stickers to label each container of prepared buffer solution.
Customers Who Bought This Also Bought