Diamond® RealSeal™ Bottles, Narrow Mouth, Low Density Polyethylene, Globe Scientific
Catalog #: 76360-374
Supplier: Globe Scientific
Specifications
- Cap Size:53 mm
- Capacity:8 L
- Ø:195 mm
- Cat. no.:76360-374
- Neck I.D.:39 mm
- Bottle Style:Round
- Height with Cap:415 mm
Specifications
About this item
Diamond® RealSeal™ Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.
- Better clarity than HDPE
- Flexible with excellent impact resistance
- Medical grade resins meet demanding requirements
- Leakproof, linerless caps for secure closing
- Laboratory bottle quality and performance with savings associated with economy bottles
- Safe, shatter-resistant alternative to glass bottles
Diamond® RealSeal™ Bottles are available in a wide range of shapes, sizes, and materials to accommodate most application needs. Small or large, wide mouth or narrow, natural or amber - even large format with handles - the Diamond® RealSeal™ line includes the most frequently used and less common bottles.
These narrow mouth bottles improve pouring of liquid contents making them perfect for storing and shipping liquids.
Customers Who Bought This Also Bought