Order Entry
ContactUsLinkComponent
Bottle Wide Mouth Round Pp 60 ml PK12
Diamond® RealSeal™ Bottles, Wide Mouth, Polypropylene, Globe Scientific
Catalog #: 76360-278
Supplier:  Globe Scientific
Bottle Wide Mouth Round Pp 60 ml PK12
Diamond® RealSeal™ Bottles, Wide Mouth, Polypropylene, Globe Scientific
Catalog #: 76360-278
Supplier:  Globe Scientific

Specifications

  • Cap Size:
    28 mm
  • Capacity:
    60 ml
  • Ø:
    39 mm
  • Cat. no.:
    76360-278
  • Neck I.D.:
    21 mm
  • Bottle Style:
    Round
  • Height with Cap:
    85 mm

Specifications

About this item

Diamond® RealSeal™ Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.

  • Suitable for autoclaving at 121 °C (including cap)
  • Improved chemical resistance and clarity
  • Resistant to most corrosives and suitable for a wide variety of laboratory uses
  • Approved for use from –40 to 121 °C
  • Medical grade resins meet demanding requirements
  • Laboratory bottle quality and performance with savings associated with economy bottles
  • Safe, shatter-resistant alternative to glass bottles

Diamond® RealSeal™ Bottles are available in a wide range of shapes, sizes, and materials to accommodate most application needs. Small or large, wide mouth or narrow, natural or amber - even large format with handles - the Diamond® RealSeal™ line includes the most frequently used and less common bottles.

These wide mouth bottles are perfect for sample collection and facilitate rapid filling and emptying of solids, powders, and viscous liquids.

Customers Who Bought This Also Bought