Dual-Tested Autoclave Biohazardous Waste Bags, Medegen Medical Products
Catalog #: 76359-166
Supplier: Medegen Medical Products, LLC
Specifications
- Capacity:13 - 16 gal.
- Dimensions:25×35"
- Color:Red
- Description:Dual tested autoclave bags
- Thickness:2.0 mil
- Cat. no.:76359-166
Specifications
About this item
Dual tested autoclave bags are built to withstand the rigors of steam sterilization while meeting the essential requirements for the collection, storage, transport, and processing of autoclavable biohazardous waste.
- Biohazardous labeling in English and Spanish
- Twist ties included for easy closure
- Made in the USA
Polypropylene film has an operating temperature of 274 °F (134 °C).
Certifications: Bags meet both ASTM D1922 (Elmendorf Tear 480g) and ASTM D1709 Dart Impact (160g) requirements. They comply with US DOT 40CFR Sections 173 -197 and regulations in all 50 states including CA, FL, and PA.
Customers Who Bought This Also Bought