VWR® 12-Channel Reagent Reservoirs
Catalog #: 76358-584
Supplier: VWR International
Specifications
- Capacity:12 × 5 ml
- Length:127 mm (5")
- Sterility:Sterile
- Width:54 mm (2 1/8")
- Description:12-channel reagent reservoir
- Height:29 mm (1 1/8")
- Cat. no.:76358-584
Specifications
About this item
These 12-channel reservoirs are made from high-quality, chemical resistant PP.
- 12 individual channels
- Each channel holds 5ml of solution and is individually numbered
- Designed for complete sample withdrawal
- Individually packaged
- Have tabs on the bottom to ensure secure stackability
- Includes a fitted lid
The reservoirs are ideal for all liquid handling applications. The rigid design of this product assures consistency in an automated environment. Each channel holds 5 ml of solution and is designed w/ U shaped channels to maximize complete sample withdrawal.
Ordering information: Available sterile and non sterile.
Customers Who Bought This Also Bought