Pm Kit 7764-71000 Std Ovrhd Drive Adap
Catalog #: 76358-536
Supplier: Bellco Glass
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
$314.91
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
$66.85 to $318.03
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$194.14 to $694.12