Flask Shake 4 Bottom Baffles 500 Ml CS12
Catalog #: 76357-488
Supplier: Bellco Glass
Customers Who Bought This Also Bought
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$146.85 to $576.51
Dichloromethane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$285.68 to $3,428.09