Flask Graduated Media with Lip 4 L CS3
Catalog #: 76357-408
Supplier: Bellco Glass
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
$175.32 to $1,620.42
WEBCOL® Alcohol Preps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$15.59 to $18.12
CrossLab Programmable Temperature Vaporizing (PTV) Inlet for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
$76.85
Choline chloride 98.0-100.5% (by anhydrous basis), BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$149.28