Vessel Sparger Rnd Btm Metal Stone 15L
Catalog #: 76356-338
Supplier: Bellco Glass
Customers Who Bought This Also Bought
Sodium dihydrogen phosphate monohydrate 98.0-102.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
$213.42 to $1,903.90
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$39.55