Bag Opener Stomacher Polymer 15.5X27Cm
Catalog #: 76355-712
Supplier: Seward Laboratory System
Customers Who Bought This Also Bought
Hexane (mixture of isomers) ≥98.5% ACS for HPLC, Supelco®
Supplier: MilliporeSigma
$649.44 to $2,496.86
Potassium dihydrogen phosphate ≥99% (by acidimetry), GR ACS, Supelco®
Supplier: MilliporeSigma
$1,033.55 to $4,052.62
n-Heptane ≥99% (n-heptane) [≥99.9% (saturated C7 hydrocarbons)], OmniSolv®, Supelco®
Supplier: MilliporeSigma
$971.75 to $3,463.97
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Extran® 300 Liquid Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
$363.65 to $1,161.45
Traceable® Certified Reference Material (CRM) pH Buffers, Control Company
Supplier: Argos Technologies
$62.31