VWR® PCR® Cooler, 96-Well
Catalog #: 76343-418
Supplier: VWR International
Specifications
- Color:Light blue/dark blue
- Description:VWR® PCR® Cooler, 96-Well, for 0.2 / 0.5 ml Tubes and Assay Plates
- Cat. no.:76343-418
Specifications
About this item
The VWR® PCR® Cooler is ideal for thermosensitive applications, PCR®, or cell-based assays, as well as gentle sample thawing and stopping of reactions.
- Modern patented design cools faster in freezers and stays cold longer on the bench top
- PCR® Cooler maintains a sample temperature of 0 °C (with 2 hours pre-cooling at -20 °C) for over an hour
- When the temperature ceiling of 6 °C has been exceeded, users are signaled by a contrasting color change of the cooler, or even a single well
- Cools industry standard format 0.2 ml and 0.5 ml PCR® tubes, strips and single format, as well as most 96 well plates, in both standard and low profile well shape
- Better alternative than ice baths as dry incubation reduces the risk of contamination
- Freeze cooler in an upright position enabling cold storage of tubes
- Well design prohibits the freezing of samples
- Fully sealed and sonic welded cooler keeps coolant contained offering worry free usage
- Save space with compact footprint and stacking ability
- The lid is ambidextrous; it can be switched to accommodate left or right-hand orientation and is detachable
Perfect for transporting samples and reactions between preparation, thermal cycling and post-PCR® procedure areas.
Material: Polypropylene (Cooler); Polycarbonate (Lid).
Customers Who Bought This Also Bought