AlphaTec® 53-001 Series Gloves, Ansell
Catalog #: 76339-858
Supplier: Ansell Healthcare
Specifications
- Ambidextrous/Hand-specific:Hand-specific
- Glove Color:Gray,Black
- Glove size:9
- Length:350 mm (13")
- Latex/Latex-free:Latex-free
- Thickness:15 mil Shell, 2 mil Grip
- Cuff style:Gauntlet
- Liner material:Nylon
- Finish:Ansell GRIP™ Technology
- Cat. no.:76339-858
Specifications
About this item
One glove with the benefits of two, offering multi-barrier chemical protection.
- Innovative multi-layer polymer design: nitrile/neoprene/nitrile
- Provides protection against a wide range of chemicals, from acids and bases to hydrocarbons and organic solvents
- Tested according to EN ISO 374:2016; permeation time >30 minutes against 13 chemicals out of 18 listed
- Features the latest MICROCHEM™ chemical barrier technology, providing superior protection for use in hazardous environments
- ANSELL GRIP™ technology provides enhanced dexterity, grip, and comfort when handling wet or oily items
- Inner soft nylon liner for comfort and increased mechanical protection
The Ansell AlphaTec® 53-001 is a 'two-in-one' nitrile and neoprene multilayer glove that provides broader chemical protection, helps reduce the number of SKUs used in a workplace, and ensures peace of mind for safety managers and workers in hazardous chemical applications.
Certifications: EN388:2016 3121A; EN407 X1XXXX; EN420:2003 + A1:2009; EN ISO 374-1:2016 Type A ACFGJKLMNOPST.
Packaging: Gloves are packaged 6 pairs per bag, 12 bags per carton.
Customers Who Bought This Also Bought