Order Entry
ContactUsLinkComponent
Standard Conductivity/Tds 111900Us 300Ml
Oakton® Conductivity and TDS Standards, Cole Parmer
Catalog #: 76338-554
Supplier:  Argos Technologies
CAS Number:  
Standard Conductivity/Tds 111900Us 300Ml
Oakton® Conductivity and TDS Standards, Cole Parmer
Catalog #: 76338-554
Supplier:  Argos Technologies
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Conductivity:
    111900 µS/cm
  • Description:
    Conductivity and TDS standard
  • Size:
    300 ml (5×60 ml)
  • TDS calibration:
    -
  • TDS conversion factor:
    -
  • Cat. no.:
    76338-554

Specifications

About this item

Ideal for laboratory and field analysis.

  • Traceable to NIST standard reference materials
  • Solutions help you calibrate conductivity instruments to ensure accuracy
  • Standard values given at 25 °C

Oakton conducitivity and TDS standards feature temperature corrections, KCl, NaCL, and 442 conversions printed directly on the bottle for ease of use (excluded on 60-mL bottle, but included in insert).

Certifications: These standards are manufactured under ISO 9000 quality guidelines and traceable to NIST standards.
Caution: If unopened, shelf life is one year. If opened, shelf life if 4 months.
Customers Who Bought This Also Bought