To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Conductivity:1413 µS/cm
- Description:Conductivity and TDS standard
- Size:300 ml (5×60 ml)
- TDS calibration:744.7 (ppm KCl)
702.1 (ppm NaCl) - TDS conversion factor:1000 (442 ppm)
- Cat. no.:76338-550
Specifications
About this item
Ideal for laboratory and field analysis.
- Traceable to NIST standard reference materials
- Solutions help you calibrate conductivity instruments to ensure accuracy
- Standard values given at 25 °C
Oakton conducitivity and TDS standards feature temperature corrections, KCl, NaCL, and 442 conversions printed directly on the bottle for ease of use (excluded on 60-mL bottle, but included in insert).