Empore™ Sealing Tape Pads, CDS Analytical
Catalog #: 76333-160
Supplier: CDS ANALYTICAL
Specifications
- Description:Empore™ sealing tape pad
- Cat. no.:76333-160
Specifications
About this item
Empore™ sealing tape pads are ideal for covering a 96-well plate during storage or transfer to another work station. They are not intended for use with auto-injectors.
Empore™ tape pads are intended for sealing unused portions of a 96-well solid phase extraction plate during vacuum processing to enable vacuum to be uniformly applied to each well being used. Use a clean sheet each time for maximum sealing ability.
Caution: These tape sheets are not intended for sealing collection plates prior to sample injection because the adhesive may clog the injection needle.
Customers Who Bought This Also Bought